Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human THBS1 (P07996) native Protein from human platelets

Catalog No. 89964635 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 ug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-635 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-964-635 Supplier Abnova™ Supplier No. P4261

missing translation for 'validMainframeMsg'

Used for SDS-PAGE

The protein encoded by this gene is a subunit of a disulfide-linked homotrimeric protein. This protein is an adhesive glycoprotein that mediates cell-to-cell and cell-to-matrix interactions. This protein can bind to fibrinogen, fibronectin, laminin, type V collagen and integrins alpha-V/beta-1. This protein has been shown to play roles in platelet aggregation, angiogenesis, and tumorigenesis. [provided by RefSeq]

Specifications

Accession Number P07996
For Use With (Application) SDS-PAGE
Formulation Lyophilized from 20mM Tris-HCl, 600mM NaCl, 2mM CaCl
Gene ID (Entrez) 7057
Molecular Weight (g/mol) 420kDa
Name THBS1 (Human) Native Protein
Preparation Method Native protein purified from human platelets
Quantity 25 ug
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with deionized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias THBS/TSP/TSP1
Common Name THBS1
Gene Symbol THBS1
Species Human
Protein Tag None
Expression System Native protein purified from human platelets
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.