Learn More
Biomatik Corporation Human Thioredoxin 2, Mitochondrial (TXN2) ELISA Kit
Supplier:  Biomatik Corporation EKU11740
Encompass_Preferred
Human Thioredoxin 2, Mitochondrial (TXN2) ELISA Kit for detecting TXN2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99757. Gene ID: 25828. Target Synonyms: MT-TRX; TRX2; MTRX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5003