Learn More
Sino Biological Anti-TPO/Thrombopoietin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10381T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Thrombopoietin / THPO (rh Thrombopoietin / THPO; Catalog#10381-H07H; NP_000451.1; Ser22-Gly353). Thrombopoietin / THPO specific IgG was purified by Human Thrombopoietin / THPO affinity chromatography.
Catalog No. 50-240-9301