Learn More
Sino Biological Recombinant Human TPO/Thrombopoietin Protein (His Tag)
Supplier:  Sino Biological 10381H07H1
Encompass_Preferred
A DNA sequence encoding the mature form of human THPO (NP_000451.1) (Ser 22-Gly 353) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-5345