Learn More
Sino Biological Human THPO Protein 1MG (Sino Biological)
Supplier:  Sino Biological 10381H08H11
Encompass_Preferred
A DNA sequence encoding the human THPO (NP_000451.1) (Met1-Leu192) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3115