Learn More
Bioworld HUMAN THREONYL T-RNA SYNTHE
Supplier:  Bioworld 220604931
Encompass_Preferred
HUMAN THREONYL T-RNA SYNTHETASE (RECOMBINANT): Recombinant Human Threonyl-tRNA synthetase (PL-7) Source:Sf9 insect cells. Background: Threonyl-tRNA synthetase is a member of the aminoacyl-tRNA synthetase family, key enzymes of protei...
Catalog No. 50-253-588