Learn More
Cusabio Technology LLC Human Thrombin ELISA Kit
Catalog No. 5000051097
Encompass_Preferred
The Human Thrombin ELISA Kit is designed to quantitatively measure thrombin levels in human serum and plasma samples using a sandwich assay methodology This kit detects thrombin within a broad dynamic range of 7 8 ng/mL to 500 ng/mL with a sensitivity of 1 95 ng/mL ensuring accurate detection of low-concentration analytes The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours of processing Measurements are performed at a detection wavelength of 450 nm providing reliable and reproducible data for clinical or research applications Suitable for studies involving coagulation pathways thrombosis or hemostatic disorders this kit offers a robust tool for precise thrombin quantification in biological fluids
Catalog No. 50-000-51097 Supplier Cusabio Technology LLC Supplier No. CSBE14235H
The Human Thrombin ELISA Kit is designed to quantitatively measure thrombin levels in human serum and plasma samples using a sandwich assay methodology This kit detects thrombin within a broad dynamic range of 7 8 ng/mL to 500 ng/mL with a sensitivity of 1 95 ng/mL ensuring accurate detection of low-concentration analytes The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours of processing Measurements are performed at a detection wavelength of 450 nm providing reliable and reproducible data for clinical or research applications Suitable for studies involving coagulation pathways thrombosis or hemostatic disorders this kit offers a robust tool for precise thrombin quantification in biological fluids