Learn More
ACROBiosystems Human Thrombopoietin R, His Tag (THR-H52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Trp 491 (Accession # P40238-1).
Catalog No. 502230806
Encompass_Preferred
Thrombopoietin R, also known as TPO-R, is expressed predominantly on the surface of MKs, platelets, hemangioblasts, and hematopoietic stem cells (HSCs). Binding of TPO to the megakaryocyte TPO-R leads to different effects: prevention of megakaryocyte apoptosis; increased megakaryocyte number, size, and ploidy; increasing rate of megakaryocyte maturation; and internalization of the TPO/TPO-R complex. Thrombopoietin R involved in multiple signal transduction pathways, such as JAK, STAT, and MAP kinase.
This item is not returnable. View return policy
Catalog No. 50-223-0806 Supplier ACROBiosystems Supplier No. THRH52H7100UG
Thrombopoietin R, also known as TPO-R, is expressed predominantly on the surface of MKs, platelets, hemangioblasts, and hematopoietic stem cells (HSCs). Binding of TPO to the megakaryocyte TPO-R leads to different effects: prevention of megakaryocyte apoptosis; increased megakaryocyte number, size, and ploidy; increasing rate of megakaryocyte maturation; and internalization of the TPO/TPO-R complex. Thrombopoietin R involved in multiple signal transduction pathways, such as JAK, STAT, and MAP kinase.