Learn More
Biomatik Corporation Human Thrombopoietin (TPO) CLIA Kit
Supplier:  Biomatik Corporation EKU08845
Encompass_Preferred
Human Thrombopoietin (TPO) CLIA Kit for detecting TPO in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P40225. Gene ID: 7066. Target Synonyms: THPO; MGDF; MKCSF; ML; MPLLG; Myeloproliferative Leukemia Virus Oncogene Ligand; Megakaryocyte Growth And Development Factor; Megakaryocyte Stimulating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5252