Learn More
Raybiotech Inc Human Thrombospondin 1 ELISA Kit
Supplier:  Raybiotech Inc ELHTSP11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 6 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Thrombospondin-1. It shows less than 5% cross-reactivity with recombinant human Thrombospondin-2 and recombinant human Thrombospondin-4.
Catalog No. 50-194-9826