Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Thrombospondin-5/COMP ELISA Kit

Catalog No. EH453RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH453RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH453RBX10 Supplier Invitrogen™ Supplier No. EH453RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human Thrombospondin-5/COMP quantitates human Thrombospondin-5/COMP in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Thrombospondin-5/COMP.

COMP is a noncollagenous extracellular matrix protein. It consists of five identical glycoprotein subunits, each with EGF like and calcium binding (thrombospondin like) domains. Oligomerization results from formation of a five stranded coiled coil and disulfides. Binding to other ECM proteins such as collagen appears to depend on divalent cations. Mutations can cause the osteochondrodysplasias pseudochondroplasia (PSACH) and multiple epiphyseal dysplasia (MED).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49747
Assay Range 0.085-20 ng/mL
Assay Sensitivity 0.085 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Cartilage oligomeric matrix protein; cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia 1, multiple); cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia 1, multiple); Comp; EDM1; EPD1; MED; MGC131819; MGC149768; PSACH; pseudoachondroplasia (epiphyseal dysplasia 1, multiple); putative cartilage oligomeric matrix protein; THBS5; Thrombospondin5; Thrombospondin-5; TSP 5; TSP5Cartilage oligomeric matrix protein; cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia 1, multiple); cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia 1, multiple); Comp; EDM1; EPD1; MED; MGC131819; MGC149768; PSACH; pseudoachondroplasia (epiphyseal dysplasia 1, multiple); putative cartilage oligomeric matrix protein; THBS5; Thrombospondin5; Thrombospondin-5; TSP 5; TSP5;
Gene ID (Entrez) 1311
Gene Symbol COMP
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target COMP
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.