Learn More
Bioworld Human Thrombospondin 1 (Recombinant), 2 μg
Supplier:  Bioworld 220604961
Encompass_Preferred
Synonym: Thrombospondin-1, THBS1, TSP, TSP1, THBS, THBS-1. Description: Recombinant Human THBS1 is glycosylated with N-linked sugars and produced using baculovirus vectors in insect cells. Recombinant Human THBS1 is Reactive with A4.1 anti-TSP mAb and its Mw is 140,000 Dalton. Source: Baculovirus Insect Cells. Physical appearance: Sterile Filtered clear solution. Formulation: The sterile protein solution contains 20mM Sodium phosphate, pH 6.0 and 300mM NaCl.Stability: Recombinant Human TSP should be stored between 2°- 8°C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA).Purity: Greater than 90.0% as determined by-(a) Analysis by RP-HPLC.(b) Analysis by SDS-PAGE. Usage: For Laboratory Research Use Only.
Catalog No. 50-253-591