Learn More
BIOHIPPO Human THRSP Microsample
SDP
Catalog No. 5000313771
Encompass
Human THRSP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of THRSP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human THRSP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human THRSP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human THRSP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13771 Supplier BIOHIPPO Supplier No. BHE15211599
Human THRSP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of THRSP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human THRSP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human THRSP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human THRSP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color