Learn More
Sino Biological Anti-Thy1/CD90 Antibody (PerCP), Mouse Monoclonal
Supplier:  Sino Biological 16897MM10C100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Thy1 (rh Thy1; Catalog#16897-HCCH; NP_006279.2; Met1-Cys130) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-240-9866