Learn More
Sino Biological Human THY1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 16897MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD90/THY-1 (rh CD90/THY-1; Catalog#16897-HCCH; NP_006279.2; Met1-Cys130). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2540