Learn More
Biomatik Corporation Human Thymosin Beta 10 (TMSb10) ELISA Kit
Supplier:  Biomatik Corporation EKU07665
Encompass_Preferred
Human Thymosin Beta 10 (TMSb10) ELISA Kit for detecting TMSb10 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 24.69-2000ng/ml. Sensitivity: 13.52ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P63313. Gene ID: 9168. Target Synonyms: MIG12; TB10; PTMB10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7789