Learn More
Biomatik Corporation Human Thymosin Beta 4, Y-Chromosomal (Tb4Y) ELISA Kit
Supplier:  Biomatik Corporation EKU07667
Encompass_Preferred
Human Thymosin Beta 4, Y-Chromosomal (Tb4Y) ELISA Kit for detecting Tb4Y in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 123.46-10000ng/ml. Sensitivity: 42.2ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: O14604. Gene ID: 9087. Target Synonyms: TMSB4Y; Thymosin,Beta 4,Y-Linked. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7407