Learn More
Cell Sciences Inc Native Human Thyroglobulin, Purified from Human Thyroid, Essential Research Reagent, 10 mg
Supplier:  Cell Sciences Inc CSI14829B
Encompass
Human Thyroglobulin is a critical enzyme for thyroid hormone biosynthesis and storage. Purified from human thyroid, this native glycoprotein is vital for researching thyroid function and related endocrine processes. With a molecular weight of 660 kDa and ≥96% purity, it provides reliable performance in studies. The lyophilized format enhances stability and ease of use while stored at 2-8°C.
Catalog No. NC1184784