Learn More
Biomatik Corporation Human Thyroglobulin (TG) CLIA Kit
Supplier:  Biomatik Corporation EKU08864
Encompass_Preferred
Human Thyroglobulin (TG) CLIA Kit for detecting TG in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 68.6-50,000pg/ml. Sensitivity: 23.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01266. Gene ID: 7038. Target Synonyms: AITD3; TGN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4453