Learn More
Biomatik Corporation Human Thyroid Hormone Receptor Alpha (THRa) ELISA Kit
Supplier:  Biomatik Corporation EKU07675
Encompass_Preferred
Human Thyroid Hormone Receptor Alpha (THRa) ELISA Kit for detecting THRa in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.50-4000pg/ml. Sensitivity: 22.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10827. Gene ID: 7067. Target Synonyms: THR-A; NR1A1; AR7; EAR7; ERB-T1; ERBA1; NR1-A1; THRA1; THRA2; c-ERBA1; Erythroblastic Leukemia Viral Oncogene Homolog; Nuclear Receptor Subfamily 1,Group A,Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5904