Learn More
Biomatik Corporation Human Thyroid Hormone Receptor Interactor 6 (TRIP6) ELISA Kit
Supplier:  Biomatik Corporation EKU12379
Encompass_Preferred
Human Thyroid Hormone Receptor Interactor 6 (TRIP6) ELISA Kit for detecting TRIP6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15654. Gene ID: 7205. Target Synonyms: OIP1; ZRP-1; Opa-interacting protein 1; Zyxin-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5134