Learn More
Biomatik Corporation Human Thyroid Hormone Responsive (THRSP) CLIA Kit
Supplier:  Biomatik Corporation EKU09856
Encompass_Preferred
Human Thyroid Hormone Responsive (THRSP) CLIA Kit for detecting THRSP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q92748. Gene ID: 7069. Target Synonyms: S14; SPOT14; LPGP1; Lipogenic Protein 1; Thyroid hormone-inducible hepatic protein; Spot 14 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6345