Learn More
Raybiotech Inc Human Thyroid Peroxidase ELISA Kit
Supplier:  Raybiotech Inc ELHTPX1
Encompass_Preferred
For cell culture supernatants, EDTA treated plasma, and serum samples. Heparin and citrate are not recommended. Sensitivity: 0.3 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human TPX. Other species not determined.
Catalog No. 50-194-9802