Learn More
Sino Biological Human Thyroid peroxidase/TPO Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785107
Encompass_Preferred
The Human Thyroid peroxidase / TPO ELISA Pair Set is for the quantitative determination of Human Thyroid peroxidase / TPO.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Thyroid peroxidase / TPO coated on a 96-well plate. Standards and samples are added to the wells, and any Human Thyroid peroxidase / TPO present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human Thyroid peroxidase / TPO monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Thyroid peroxidase / TPO present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85107 Supplier Sino Biological Supplier No. SEK1319415
The Human Thyroid peroxidase / TPO ELISA Pair Set is for the quantitative determination of Human Thyroid peroxidase / TPO.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Thyroid peroxidase / TPO coated on a 96-well plate. Standards and samples are added to the wells, and any Human Thyroid peroxidase / TPO present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human Thyroid peroxidase / TPO monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Thyroid peroxidase / TPO present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.