Learn More
Biomatik Corporation Human Thyroxine Binding Globulin (TBG) CLIA Kit
Supplier:  Biomatik Corporation EKU08863
Encompass_Preferred
Human Thyroxine Binding Globulin (TBG) CLIA Kit for detecting TBG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.14-100ng/ml. Sensitivity: 0.07ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P05543. Gene ID: 6906. Target Synonyms: SERPINA7; Serpin Peptidase Inhibitor Clade A Member 7; T4-binding globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5274