Learn More
Sino Biological Human Tie-1 Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK105095
Encompass_Preferred
The Human Tie1 ELISA Pair Set is for the quantitative determination of Human Tie1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Tie1 coated on a 96-well plate. Standards and samples are added to the wells, and any Tie1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Tie1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Tie1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85233