Learn More
Sino Biological Anti-Tie-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10509RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Tie1 (rh Tie1; Catalog#10509-H08H; NP_005415.1; Met 1-Gln 760). Tie1 specific IgG was purified by Human Tie1 affinity chromatography.
Catalog No. 50-241-1168