Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TIGAR Recombinant Protein, PeproTech®

Catalog No. p-7246328
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15014100UG 100 μg
15-014-01M 2 x 500 μg
15014250UG 250 μg
150-14-25UG 25 μg
15014500UG 500 μg
150-14-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 15014100UG Supplier Gibco™ Supplier No. 15014100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

150-14-1MG will be provided as 2 x 500 μg (150-14-500UG). Recombinant Human TIGAR expressed in E.coli is a 29.9 kDa protein containing 269 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene encodes a member of the kelch family of proteins, which are characterized by kelch repeat motifs and a POZ/BTB protein-binding domain. It is thought that kelch repeats are actin binding domains. However, the specific function of this protein has not been determined. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NQ88
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 57103
Molecular Weight (g/mol) 29.9 kDa
Name Human TIGAR
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 9630033F20Rik; AA793651; AI595337; C12orf5; C27H12orf5; C79710; C85509; FR2BP; Fructose-2,6-bisphosphatase TIGAR; fructose-2,6-bisphosphate 2-phosphatase; hypothetical protein LOC502894; LOC502894; probable fructose-2,6-bisphosphatase TIGAR; TIGAR; TP53 induced glycolysis regulatory phosphatase; TP53-induced glycolysis and apoptosis regulator; TP53-induced glycolysis regulatory phosphatase; transactivated by NS3TP2 protein; Trp53 induced glycolysis repulatory phosphatase
Common Name TIGAR
Gene Symbol TIGAR
Conjugate Unconjugated
Recombinant Recombinant
Sequence ARFALTVVRH GETRFNKEKI IQGQGVDEPL SETGFKQAAA AGIFLNNVKF THAFSSDLMR TKQTMHGILE RSKFCKDMTV KYDSRLRERK YGVVEGKALS ELRAMAKAAR EECPVFTPPG GETLDQVKMR GIDFFEFLCQ LILKEADQKE QFSQGSPSNC LETSLAEIFP LGKNHSSKVN SDSGIPGLAA SVLVVSHGAY MRSLFDYFLT DLKCSLPATL SRSELMSVTP NTGMSLFIIN FEEGREVKPT VQCICMNLQD HLNGLTETR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.