Learn More
Biolegend Ultra-LEAF™ Purified anti-human TIGIT (VSTM3)
Supplier:  Biolegend 372720
Encompass_Preferred
Ultra-LEAF™ Purified anti-human TIGIT (VSTM3) [A15153G]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 1 mg
Catalog No. NC1319203