Learn More
Abcam Human TIGIT ELISA Kit - Intracellular, 1 x 96 Tests
Catalog No. 502699109
Encompass_Preferred
Human TIGIT ELISA Kit - Intracellular is a single-wash 90-min Simplestep used to quantify Human TIGIT with a sensitivity of 4 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-9109 Supplier Abcam Supplier No. AB3145981X96TESTS
Human TIGIT ELISA Kit - Intracellular is a single-wash 90-min Simplestep used to quantify Human TIGIT with a sensitivity of 4 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement