Learn More
Sino Biological Human TIGIT MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10917MM5220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TIGIT (rh TIGIT; Catalog#10917-H08H; NP_776160.2; Met1-Phe138). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1182