Learn More
Sino Biological Human TIGIT Protein 500ug (Sino Biological)
Supplier:  Sino Biological 10917H08H3500
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human TIGIT (NP_776160.2) (Met1-Pro141) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-237-0739