Learn More
Sino Biological Recombinant Human TIGIT Protein (Dimer, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10917H49H1B20
Encompass_Preferred
A DNA sequence encoding the Human TIGIT (NP_776160.2) (Met22-Pro141) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed. This construct has a dimer design.
Catalog No. 50-255-5072