Learn More
Sino Biological Recombinant Human TIGIT Protein (ECD, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 10917H49HB20
Encompass_Preferred
A DNA sequence encoding the human TIGIT (Q495A1-1)(Met1-Pro141) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-224-6726