Learn More
Bon Opus Biosciences Recombinant Human Tim-1 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP103350UG
Encompass_Preferred
Recombinant Human Tim-1 (C-hFc) is produced by Human Cells. The target gene encoding Ser21-Gly295 is expressed with a C-hFc tag. Accession# Q96D42. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1033
Catalog No. 50-217-2632