Learn More
Bon Opus Biosciences Recombinant Human Tim-1 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP103450UG
Encompass_Preferred
Recombinant Human Tim-1 (C-mFc) is produced by Human Cells. The target gene encoding Ser21-Gly295 is expressed with a C-mFc tag. Accession# Q96D42. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1034
Catalog No. 50-217-2633