Learn More
Raybiotech Inc Human TIM-3 (KIM-3) ELISA Kit
Supplier:  Raybiotech Inc ELHTIM31
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 10 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human T Cell Immunoglobulin Mucin-3 (HAVCR2 / KIM-3)
Catalog No. 50-194-9781