Learn More
Sino Biological Human TIM-3/HAVCR2 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786730
Encompass_Preferred
The Human TIM-3/HAVCR2 ELISA Pair Set is for the quantitative determination of Human TIM-3/HAVCR2.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human TIM-3/HAVCR2 coated on a 96-well plate. Standards and samples are added to the wells, and any Human TIM-3/HAVCR2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human TIM-3/HAVCR2 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human TIM-3/HAVCR2 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86730 Supplier Sino Biological Supplier No. SEK103905
The Human TIM-3/HAVCR2 ELISA Pair Set is for the quantitative determination of Human TIM-3/HAVCR2.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human TIM-3/HAVCR2 coated on a 96-well plate. Standards and samples are added to the wells, and any Human TIM-3/HAVCR2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human TIM-3/HAVCR2 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human TIM-3/HAVCR2 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.