Learn More
Sino Biological Recombinant Human TIM-3/HAVCR2 Protein (ECD, hFc Tag) 1 mg
Supplier:  Sino Biological 10390H02H1
Encompass_Preferred
A DNA sequence encoding the human HAVCR2 (NP_116171.3) (Met1-Arg200) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6983