Learn More
Sino Biological Recombinant Human TIM-3/HAVCR2 Protein (hFc & AVI Tag), Biotinylated
Supplier:  Sino Biological 10390H41HB20
Encompass_Preferred
A DNA sequence encoding the human HAVCR2 (NP_116171.3)(Met1-Arg200) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-224-7277