Learn More
Sino Biological Human TIMD4 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 12161T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TIMD4 (rh TIMD4; Catalog#12161-H08H; AAH08988.1; Met1-Leu315). TIMD4 specific IgG was purified by Human TIMD4 affinity chromatography.
Catalog No. 50-237-1581