Learn More
Sino Biological Recombinant Human Tim4/TIMD4 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 12161H08HB20
Encompass_Preferred
A DNA sequence encoding the human TIMD4 (AAH08988.1) extracellular domain (Met 1-Leu 315) was expressed, fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4450