Learn More
Raybiotech Inc Human TIMP-1 ELISA Kit
Supplier:  Raybiotech Inc ELHTIMP11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 40 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human ANG, CD23, Eotaxin, GCSF, GM-CSF, GRO-alpha, GRO-beta, GRO-gamma, I-309, IFN-gamma, IL-1 alpha, IL-1 beta, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p40), IL-12 (p70), IL-15, IL-16, IP-10, MCP-1, MCP-2, MCP-3, MCP-4, MCSF, MIG, MIP-1 alpha, MIP-1 beta, NAP-2, PDGF, PF-4, PARC, SCF, SDF-1 alpha, TNFbeta, TGFbeta1, TGFbeta2, TGFbeta3, VEGF.
Catalog No. 50-194-9783