Learn More
Sino Biological Anti-TIMP-1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10934MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TIMP1 (rhTIMP1; Catalog#10934-H08H; NP_003245.1; Met 1-Ala 207). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5055