Learn More
Sino Biological Recombinant Anti-TIMP-1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10934R004100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TIMP1 (rh TIMP1; Catalog#10934-H08H; NP_003245.1; Met 1-Ala 207).
Catalog No. 50-255-5053