Learn More
Sino Biological Anti-TIMP-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10934RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TIMP1 (rhTIMP1; Catalog#10934-H08H; NP_003245.1; Met 1-Ala 207). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-5051