Learn More
Sino Biological Anti-TIMP-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10934T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TIMP-1 / TIMP1 (rh TIMP-1 / TIMP1; Catalog#10934-H08H; NP_003245.1; Met1-Ala207). TIMP-1 / TIMP1 specific IgG was purified by Human TIMP-1 / TIMP1 affinity chromatography.
Catalog No. 50-255-5050