Learn More
Biomatik Corporation Human Tissue Factor Pathway Inhibitor (TFPI) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU11537
Encompass_Preferred
Human Tissue Factor Pathway Inhibitor (TFPI) Wide-range ELISA Kit for detecting TFPI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.34ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10646. Gene ID: 7035. Target Synonyms: TFI; EPI; LACI; TFPI1; Lipoprotein-Associated Coagulation Inhibitor; Extrinsic pathway inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8128