Learn More
Biomatik Corporation Human Tissue Inhibitors Of Metalloproteinase 3 (TIMP3) CLIA Kit
Supplier:  Biomatik Corporation EKU09451
Encompass_Preferred
Human Tissue Inhibitors Of Metalloproteinase 3 (TIMP3) CLIA Kit for detecting TIMP3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.08-60ng/ml. Sensitivity: 0.03ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P35625. Gene ID: 7078. Target Synonyms: SFD; TIMP Metallopeptidase Inhibitor 3; Metallopeptidase Inhibitor 3; Sorsby Fundus Dystrophy; Pseudoinflammatory. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6967