Learn More
Biomatik Corporation Human Tissue Inhibitors Of Metalloproteinase 1 (TIMP1) CLIA Kit
Supplier:  Biomatik Corporation EKU08888
Encompass_Preferred
Human Tissue Inhibitors Of Metalloproteinase 1 (TIMP1) CLIA Kit for detecting TIMP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01033. Gene ID: 7076. Target Synonyms: EPO; CLGI; EPA; HCI; Erythroid Potentiating Activity; Collagenase Inhibitor; Metalloproteinase inhibitor 1; Fibroblast collagenase inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6617